• USA Home
  • Search Results

  • fmoclyszoh502568606082411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fmoc-Lys(Z)-OH

2 Products
Synonym:   N-α-Fmoc-N-ε-benzyloxycarbonyl-L-lysine
Image of Fmoc-Lys(Z)-OH
CAS Number:
Empirical Formula (Hill Notation):
 C29H30N2O6
Molecular Weight:
 502.56
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More