• USA Home
  • Search Results

  • fmoctyrtbuoh459537198938311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fmoc-Tyr(tBu)-OH

1 Products
Synonym:   N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-tyrosine, Fmoc-O-tert-butyl-L-tyrosine
Image of Fmoc-Tyr(<I>t</I>Bu)-OH
CAS Number:
Empirical Formula (Hill Notation):
 C28H29NO5
Molecular Weight:
 459.53
 Beilstein Registry Number:
 4216652
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss
 32160406
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More