• USA Home
  • Search Results

  • folicacid441405930311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Folic acid

10 Products
Synonym:   PteGlu, Pteroyl-L-glutamic acid, Vitamin M
Image of Folic acid
CAS Number:
Empirical Formula (Hill Notation):
 C19H19N7O6
Molecular Weight:
 441.40
 Beilstein/REAXYS Number:
 100781
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 34058014
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Agency/Method
  Biological Source
  Anal. Standard Form
  Assay
  Special Grade
  Titertest
  Melting Point
  Color
  Physical Form
Show More