• USA Home
  • Search Results

  • fombliny123456999167911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fomblin® Y

4 Products
Image of Fomblin<SUP>®</SUP> Y
CAS Number:
Linear Formula:
 CF3O[-CF(CF3)CF2O-]x(-CF2O-)yCF3
 MDL number:
 NACRES:
 NB.79
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Density
  Refractive Index
  Viscosity
  Molecular weight
Show More