• USA Home
  • Search Results

  • formaldehydesolution30035000011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Formaldehyde solution

7 Products
Synonym:   Formalin
Image of Formaldehyde solution
CAS Number:
Linear Formula:
 HCHO
Molecular Weight:
 30.03
 Beilstein/REAXYS Number:
 1209228
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Agency/Method
  pH Value
  UV-Absorption Values
  Quality clarifier
  Color
  Restriction
  Product Line
  Melting Point
  Suitability
  Application(s)
  Physical Form
  Special Grade
  Assay
  Additive
Show More