• USA Home
  • Search Results

  • formamide45047512711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Formamide

10 Products
Synonym:   Amide C1, Formic acid amide, Methane amide, Formic amide
Image of Formamide
CAS Number:
Linear Formula:
 HCONH2
Molecular Weight:
 45.04
 Beilstein/REAXYS Number:
 505995
 EC Index Number:
 200-842-0
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  UV-Absorption Condition
  Refractive Index
  Physical Form
  Purity Grade
  Product Line
  Ice Shipping
  Color
  Restriction
  Suitability
  Potency
  pH Value
  UV-Absorption Values
  Transition temperature
  Explosion limit
  Special Grade
Show More