• USA Home
  • Search Results

  • formicacid46036418611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Formic acid

9 Products
Image of Formic acid
CAS Number:
Linear Formula:
 HCOOH
Molecular Weight:
 46.03
 Beilstein/REAXYS Number:
 1209246
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Document Links
  Allergen
  Additive
  Solubility
  Quality clarifier
  Miscellaneous Specifications
  Color
  Agency/Method
  Purity Grade
  Assay
Show More