• USA Home
  • Search Results

  • formicacid9810046036418611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Formic acid 98% - 100%

5 Products
Synonym:   Formic acid, Methanoic acid, Formylic acid
Image of Formic acid 98% - 100%
CAS Number:
Linear Formula:
 HCOOH
Molecular Weight:
 46.03
 Beilstein/REAXYS Number:
 1209246
 EC Index Number:
 200-579-1
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Explosion limit
  Application(s)
  Special Grade
  pH Value
  Transition temperature
Show More