• USA Home
  • Search Results

  • formononetin2682648572311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Formononetin

4 Products
Synonym:   7-Hydroxy-3-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 7-Hydroxy-3-(4-methoxyphenyl)chromone, 7-Hydroxy-4′-methoxyisoflavone, Biochanin B, Daidzein 4′-methyl ether, Formonetin, Formononetol
Image of Formononetin
CAS Number:
Empirical Formula (Hill Notation):
 C16H12O4
Molecular Weight:
 268.26
 Beilstein/REAXYS Number:
 237979
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Manufacturer designation
  Special Grade
Show More