• USA Home
  • Search Results

  • formothionsolution25727254082111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Formothion solution

1 Products
Synonym:   S-[2-(Formylmethylamino)-2-oxoethyl] O,O-dimethyl dithiophosphate
Image of Formothion solution
CAS Number:
Empirical Formula (Hill Notation):
 C6H12NO4PS2
Molecular Weight:
 257.27
 Beilstein/REAXYS Number:
 5942199
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More