• USA Home
  • Search Results

  • foscarnetsodium300043415656411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Foscarnet sodium

2 Products
Synonym:   Foscarnet, Phosphonoformic acid trisodium salt hexahydrate, Sodium phosphonoformate tribasic hexahydrate
Image of Foscarnet sodium
CAS Number:
Linear Formula:
 Na2O3PCO2Na · 6H2O
Molecular Weight:
 300.04
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Manufacturer designation
Show More