• USA Home
  • Search Results

  • fructose180165748711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fructose

3 Products
Synonym:   D-(−)-Fructose, D-Levulose, Fruit sugar
Image of Fructose
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O6
Molecular Weight:
 180.16
 Beilstein/REAXYS Number:
 1239004
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Manufacturer designation
  Anal. Standard Form
  Physical Form
Show More