• USA Home
  • Search Results

  • fucoxanthin65891335186811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fucoxanthin

2 Products
Synonym:   all-trans-Fucoxanthin
Image of Fucoxanthin
CAS Number:
Empirical Formula (Hill Notation):
 C42H58O6
Molecular Weight:
 658.91
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More