• USA Home
  • Search Results

  • fullerenec60720649968596811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fullerene-C60

3 Products
Synonym:   Buckminsterfullerene
Image of Fullerene-C<SUB>60</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 C60
Molecular Weight:
 720.64
 Beilstein/REAXYS Number:
 5901022
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  OLED Device Performance
  Greener Alternative Principles
  Physical Form
  Solubility
  Assay
  Orbital energy
Show More