• USA Home
  • Search Results

  • fulvestrantforsystemsuitability6067712945361811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fulvestrant for system suitability

1 Products
Synonym:   (7α,17β)-7-[9-[(4,4,5,5,5-Pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-triene-3,17-diol, Faslodex, Fulvestrant, ICI 182,780, ZD 182780, ZD 9238, ZM 182780
Image of Fulvestrant for system suitability
CAS Number:
Empirical Formula (Hill Notation):
 C32H47F5O3S
Molecular Weight:
 606.77
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More