• USA Home
  • Search Results

  • fumaricacid1160711017811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fumaric acid

14 Products
Synonym:   Fumaric acid, trans-1,2-Ethylentricarboxylic acid
Image of Fumaric acid
CAS Number:
Empirical Formula (Hill Notation):
 C4H4O4
Molecular Weight:
 116.07
 EC Index Number:
 203-743-0
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Special Grade
  Allergen
  Organoleptic property
  pH Value
  Vapor pressure
  Transition temperature
  Melting Point
  Density
  Autoignition temp
  Anal. Standard Form
  Solubility
  Purity Grade
  Product Line
  Manufacturer designation
  Biological Source
  Potency
  Agency/Method
  Application(s)
  Physical Form
  Assay
Show More

Product #
Description
47910 Fumaric acid, ≥99.0% (T),
F8509 Fumaric acid, BioReagent, suitable for cell culture,
W248800 Fumaric acid, FCC, FG,
03761 Fumaric acid, tested according to USP/NF,
76635 Fumaric acid, certified reference material, TraceCERT®,
8.17073 Fumaric acid, EMPROVE® ESSENTIAL NF,JPE,
1286708 Fumaric acid, United States Pharmacopeia (USP) Reference Standard,
ARK2164 Fumaric acid, PharmaGrade, USP/NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.,
8.00269 Fumaric acid, for synthesis,
F0600000 Fumaric acid, European Pharmacopoeia (EP) Reference Standard,
798673 Fumaric acid, anhydrous, free-flowing, Redi-Dri, ≥99%,
W248835 Fumaric acid, natural (US), ≥98%, FG,
PHL80141 Fumaric acid, phyproof® Reference Substance,
PHR1270 Fumaric Acid, Pharmaceutical Secondary Standard; Certified Reference Material,