• USA Home
  • Search Results

  • fura2am10018510896432511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fura 2-AM

2 Products
Synonym:   Fura-2 pentakis(acetoxymethyl) ester
Image of Fura 2-AM
CAS Number:
Empirical Formula (Hill Notation):
 C44H47N3O24
Molecular Weight:
 1001.85
 Beilstein/REAXYS Number:
 8183748
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
Show More