• USA Home
  • Search Results

  • furan680711000911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Furan

3 Products
Synonym:   Furfuran, Furfuran
Image of Furan
CAS Number:
Empirical Formula (Hill Notation):
 C4H4O
Molecular Weight:
 68.07
 EC Index Number
 603-105-00-5
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
  Additive
  Solubility
Show More