• USA Home
  • Search Results

  • furazolidone225166745811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Furazolidone

3 Products
Synonym:   3-(5-Nitrofurfurylideneamino)-2-oxazolidinone
Image of Furazolidone
CAS Number:
Empirical Formula (Hill Notation):
 C8H7N3O5
Molecular Weight:
 225.16
 Beilstein/REAXYS Number:
 8317414
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Mode of Action
  Physical Form
  Special Grade
  Titertest
  Solubility
Show More