• USA Home
  • Search Results

  • furfural96089801111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Furfural

7 Products
Synonym:   2-Furaldehyde, Furan-2-carboxaldehyde
Image of Furfural
CAS Number:
Empirical Formula (Hill Notation):
 C5H4O2
Molecular Weight:
 96.08
 Beilstein/REAXYS Number:
 105755
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39150703
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Vapor pressure
  Transition temperature
  Potency
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Solubility
  Physical Form
  Assay
  Agency/Method
Show More