• USA Home
  • Search Results

  • furfurylglycidylether15416538087011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Furfuryl glycidyl ether

1 Products
Synonym:   2,3-Epoxypropyl 2-furylmethyl ether, 2-[(Oxiranylmethoxy)methyl]furan
Image of Furfuryl glycidyl ether
CAS Number:
Empirical Formula (Hill Notation):
 C8H10O3
Molecular Weight:
 154.16
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More