• USA Home
  • Search Results

  • g418disulfatesalt6927110832142211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

G 418 disulfate salt

3 Products
Synonym:   Antibiotic G418
Image of G 418 disulfate salt
CAS Number:
Empirical Formula (Hill Notation):
 C20H40N4O10 · 2H2SO4
Molecular Weight:
 692.71
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
  Titertest
  Product Line
  Mode of Action
  Activity Spectrum
  Solubility
  Package quantity
  Color
  Physical Form
Show More