• USA Home
  • Search Results

  • gadoliniumiiichloride263611013852011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gadolinium(III) chloride

1 Products
Synonym:   Gadolinium trichloride
Image of Gadolinium(III) chloride
CAS Number:
Linear Formula:
 GdCl3
Molecular Weight:
 263.61
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Special Grade
Show More