• USA Home
  • Search Results

  • galangin2702454883411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Galangin

3 Products
Synonym:   3,5,7-Trihydroxyflavone, Norizalpinin
Image of Galangin
CAS Number:
Empirical Formula (Hill Notation):
 C15H10O5
Molecular Weight:
 270.24
 Beilstein/REAXYS Number:
 272179
 EC Number:
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Special Grade
  Color
Show More