• USA Home
  • Search Results

  • galaninporcine3210528881336911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Galanin porcine

1 Products
Image of Galanin porcine
CAS Number:
Empirical Formula (Hill Notation):
 C146H213N43O40
Molecular Weight:
 3210.52
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More