• USA Home
  • Search Results

  • gallicacid1701214991711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gallic acid

4 Products
Synonym:   3,4,5-Trihydroxybenzoic acid
Image of Gallic acid
CAS Number:
Linear Formula:
 (HO)3C6H2CO2H
Molecular Weight:
 170.12
 Beilstein/REAXYS Number:
 2050274
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Special Grade
Show More