• USA Home
  • Search Results

  • gallium6972744055311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gallium

5 Products
Image of Gallium
CAS Number:
Linear Formula:
 Ga
Molecular Weight:
 69.72
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 38120701
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More