• USA Home
  • Search Results

  • gaminobutyricacid103125612211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

γ-Aminobutyric acid

6 Products
Synonym:   3-Carboxypropylamine, 4-Aminobutanoic acid, GABA, Piperidic acid, Piperidinic acid, gamma-Aminobutyric acid
Image of γ-Aminobutyric acid
CAS Number:
Linear Formula:
 NH2(CH2)3COOH
Molecular Weight:
 103.12
 Beilstein/REAXYS Number:
 906818
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More