• USA Home
  • Search Results

  • garlicoil12345800078011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Garlic oil

1 Products
Synonym:   Allium sativum bulb oil
CAS Number:
 FEMA Number:
 2503
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More

Product #
Description
W250320 Garlic oil, FG,