• USA Home
  • Search Results

  • gbhc290835889911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

γ-BHC

2 Products
Synonym:   γ-1,2,3,4,5,6-Hexachlorocyclohexane, γ-benzene hexachloride, Lindane solution
CAS Number:
Empirical Formula (Hill Notation):
 C6H6Cl6
Molecular Weight:
 290.83
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More