• USA Home
  • Search Results

  • gbutyrolactone86099648011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

γ-Butyrolactone

3 Products
Synonym:   γ-Hydroxybutyric acid lactone, 4-Hydroxybutyric acid lactone, GBL
Image of γ-Butyrolactone
CAS Number:
Empirical Formula (Hill Notation):
 C4H6O2
Molecular Weight:
 86.09
 Beilstein/REAXYS Number:
 105248
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More