• USA Home
  • Search Results

  • gcarboxydlglutamicacid191145627199911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

γ-Carboxy-DL-glutamic acid

1 Products
Synonym:   3-Amino-1,1,3-propanetricarboxylic acid
Image of γ-Carboxy-<SC>DL</SC>-glutamic acid
CAS Number:
Linear Formula:
 NH2CH(COOH)CH2CH(COOH)2
Molecular Weight:
 191.14
 Beilstein/REAXYS Number:
 2214048
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More