• USA Home
  • Search Results

  • gdecalactone1702570614911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

γ-Decalactone

2 Products
Synonym:   γ-Decanolactone, 4-Hydroxydecanoic acid γ-lactone
Image of γ-Decalactone
CAS Number:
Empirical Formula (Hill Notation):
 C10H18O2
Molecular Weight:
 170.25
 Council of Europe no.:
 2230
 EC Number:
 FEMA Number:
 2360
 Flavis number:
 10.017
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More