• USA Home
  • Search Results

  • gdecanolactone1702570614911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

γ-Decanolactone

2 Products
Synonym:   4-Hydroxydecanoic acid γ-lactone
Image of γ-Decanolactone
CAS Number:
Empirical Formula (Hill Notation):
 C10H18O2
Molecular Weight:
 170.25
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More