• USA Home
  • Search Results

  • gelatinfrombovineskin12345900070811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gelatin from bovine skin

3 Products
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.75
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Restriction
  Special Grade
  Package quantity
  Physical Form
  Type
  Solubility
  Biological Source
Show More