• USA Home
  • Search Results

  • gelatinfromcoldwaterfishskin12345900070811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gelatin from cold water fish skin

2 Products
Synonym:   Gelatine, Teleostean gelatin
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.75
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Additive
  Package quantity
  Biological Source
Show More