• USA Home
  • Search Results

  • gelatinfromporcineskin12345900070811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gelatin from porcine skin

11 Products
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.75
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Viscosity
  Surface coverage
  Sterilization
  Product Line
  Ice Shipping
  Shelf Life
  Assay
  Transmittance
  Quality clarifier
  Package quantity
  Loss
  pH Value
  Type
  Special Grade
  Gel Strength
  Physical Form
  Solubility
  Biological Source
Show More

Product #
Description
G1890 Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, for electrophoresis, suitable for cell culture,
G2500 Gelatin from porcine skin, gel strength 300, Type A,
48722 Gelatin from porcine skin, medium gel strength, for microbiology,
G6144 Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A,
G2625 Gelatin from porcine skin, gel strength ~175 g Bloom, Type A,
G9136 Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture,
04055 Gelatin from porcine skin, powder, for microbiology,
48724 Gelatin from porcine skin, for microbiology, high gel strength,
73865 Gelatin from porcine skin, medium gel strength, controlled endotoxin level, for microbiology,
48720 Gelatin from porcine skin, for microbiology, low gel strength,
39465 Gelatin from porcine skin, for microbiology, ultrahigh gel strength,