• USA Home
  • Search Results

  • geldanamycinfromstreptomyceshygroscopicus560643056234611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Geldanamycin from Streptomyces hygroscopicus

1 Products
Image of Geldanamycin from <I>Streptomyces hygroscopicus</I>
CAS Number:
Empirical Formula (Hill Notation):
 C29H40N2O9
Molecular Weight:
 560.64
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More