• USA Home
  • Search Results

  • gemcitabinehydrochloride2996612211103911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gemcitabine hydrochloride

4 Products
Synonym:   2′-Deoxy-2′,2′-difluorocytidine; dFdC, Gemzar (Lilly), LY-188011, dFdC, dFdCyd
Image of Gemcitabine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C9H11F2N3O4 · HCl
Molecular Weight:
 299.66
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
  Manufacturer designation
Show More