• USA Home
  • Search Results

  • geniposidicacid374342774101111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Geniposidic acid

2 Products
Synonym:   (1S,4aS,7aS)-1-(β-D-Glucopyranosyloxy)-1,4a,5,7a-tetrahydro-7-(hydroxymethyl)-cyclopenta[c]pyran-4-carboxylic acid, Geniposidic acid
Image of Geniposidic acid
CAS Number:
Empirical Formula (Hill Notation):
 C16H22O10
Molecular Weight:
 374.34
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More