• USA Home
  • Search Results

  • genistein2702444672011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Genistein

7 Products
Synonym:   4′,5,7-Trihydroxyisoflavone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one
Image of Genistein
CAS Number:
Empirical Formula (Hill Notation):
 C15H10O5
Molecular Weight:
 270.24
 Beilstein/REAXYS Number:
 263823
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Application(s)
  Melting Point
  Biological Source
  Physical Form
  Manufacturer designation
  Solubility
  Assay
Show More