• USA Home
  • Search Results

  • genistin4323852959911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Genistin

6 Products
Synonym:   4′,5,7-Trihydroxyisoflavone 7-glucoside, Genistein 7-glucoside, Genistein-7-O-β-D-glucopyranoside, Genistoside, NSC 5112
Image of Genistin
CAS Number:
Empirical Formula (Hill Notation):
 C21H20O10
Molecular Weight:
 432.38
 Beilstein/REAXYS Number:
 64479
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Application(s)
  Special Grade
  Manufacturer designation
Show More