• USA Home
  • Search Results

  • genkwanin2842643764911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Genkwanin

3 Products
Synonym:   4′,5-Dihydroxy-7-methoxyflavone, 5-Hydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-1-benzopyran-4-one, 7-O-Methylapigenin, Apigenin 7-O-methyl ether, Puddumetin
Image of Genkwanin
CAS Number:
Empirical Formula (Hill Notation):
 C16H12O5
Molecular Weight:
 284.26
 Beilstein/REAXYS Number:
 292549
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More