• USA Home
  • Search Results

  • geosminand2methylisoborneolsolution1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Geosmin and 2-Methylisoborneol Solution

2 Products
Synonym:   Drinking Water Odor Standards, Odor Standards
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More