• USA Home
  • Search Results

  • geraniol1542510624111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Geraniol

4 Products
Synonym:   trans-3,7-Dimethyl-2,6-octadien-1-ol
Image of Geraniol
CAS Number:
Linear Formula:
 (CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
Molecular Weight:
 154.25
 Beilstein/REAXYS Number:
 1722456
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Chemical Composition
  Vapor pressure
  Application(s)
  Assay
  Agency/Method
Show More