• USA Home
  • Search Results

  • geranylacetate1962910587311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Geranyl acetate

5 Products
Synonym:   trans-3,7-Dimethyl-2,6-octadien-1-yl acetate, trans-3,7-Dimethyl-2,6-octadienyl acetate
Image of Geranyl acetate
CAS Number:
Linear Formula:
 CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
Molecular Weight:
 196.29
 Council of Europe no.:
 201
 EC Number:
 FEMA Number:
 2509
 Flavis number:
 9.011
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Shelf Life
  Allergen
  Organoleptic property
  Application(s)
  Density
  Assay
  Agency/Method
Show More