• USA Home
  • Search Results

  • geranylbutyrate2243410629611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Geranyl butyrate

2 Products
Image of Geranyl butyrate
CAS Number:
Linear Formula:
 CH3CH2CH2CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
Molecular Weight:
 224.34
 Council of Europe no.:
 274c
 EC Number:
 FEMA Number:
 2512
 Flavis number:
 9.048
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Assay
  Special Grade
Show More