• USA Home
  • Search Results

  • germaniumivoxide10464131053811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Germanium(IV) oxide

4 Products
Synonym:   Germanium dioxide
Image of Germanium(IV) oxide
CAS Number:
Linear Formula:
 GeO2
Molecular Weight:
 104.64
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Density
  Solubility
  Physical Form
  Assay
Show More