• USA Home
  • Search Results

  • ginkgolidebfromginkgobilobaleaves424401529177711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ginkgolide B from Ginkgo biloba leaves

1 Products
Synonym:   BN-52021
Image of Ginkgolide B from <I>Ginkgo biloba</I> leaves
CAS Number:
Empirical Formula (Hill Notation):
 C20H24O10
Molecular Weight:
 424.40
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More