• USA Home
  • Search Results

  • glibenclamideforpeakidentification494001023821811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glibenclamide for peak identification

1 Products
Synonym:   5-Chloro-N-[4-(cyclohexylureidosulfonyl)phenethyl]-2-methoxybenzamide, Glybenclamide, Glyburide, N-p-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzenesulfonyl-N′-cyclohexylurea
Image of Glibenclamide for peak identification
CAS Number:
Empirical Formula (Hill Notation):
 C23H28ClN3O5S
Molecular Weight:
 494.00
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More